3-Amino-N-benzyl-4-chloro-N-ethylbenzenesulfonamide

Supplier: Matrix Scientific

052664-500MG 052664-2.5G
101900-542EA 364.62 USD
101900-542 101900-544
3-Amino-N-benzyl-4-chloro-N-ethylbenzenesulfonamide
3-Amino-N-benzyl-4-chloro-N-ethylbenzenesulfonamide

Hazard: Irritant

Formula: C₁₅H₁₇ClN₂O₂S
MDL Number: MFCD13383714
CAS Number: 847171-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR