2-Chloro-5-((3,4-dihydroquinolin-1(2H)-yl)sulfonyl)aniline

Supplier: Matrix Scientific

052663-500MG 052663-2.5G
101900-538EA 364.62 USD
101900-538 101900-540
2-Chloro-5-((3,4-dihydroquinolin-1(2H)-yl)sulfonyl)aniline
2-Chloro-5-((3,4-dihydroquinolin-1(2H)-yl)sulfonyl)aniline

Hazard: Irritant

Formula: C₁₅H₁₅ClN₂O₂S
CAS Number: 847171-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR