2-{4-[(3-Amino-4-chlorophenyl)sulfonyl]-1-piperazinyl}-1-ethanol

Supplier: Matrix Scientific

052660-500MG 052660-2.5G
101900-526EA 364.62 USD
101900-526 101900-528
2-{4-[(3-Amino-4-chlorophenyl)sulfonyl]-1-piperazinyl}-1-ethanol
2-{4-[(3-Amino-4-chlorophenyl)sulfonyl]-1-piperazinyl}-1-ethanol

Hazard: Irritant

Formula: C₁₂H₁₈ClN₃O₃S
MDL Number: MFCD12475154
CAS Number: 1154310-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR