3-Amino-4-chloro-N-cyclohexyl-N-ethylbenzenesulfonamide

Supplier: Matrix Scientific

052659-500MG 052659-2.5G
101900-522EA 364.62 USD
101900-522 101900-524
3-Amino-4-chloro-N-cyclohexyl-N-ethylbenzenesulfonamide
3-Amino-4-chloro-N-cyclohexyl-N-ethylbenzenesulfonamide

Hazard: Irritant

Formula: C₁₄H₂₁ClN₂O₂S
MDL Number: MFCD12440959
CAS Number: 1036541-04-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR