3-Amino-N-benzyl-4-chloro-N-methylbenzenesulfonamide

Supplier: Matrix Scientific

052658-500MG 052658-2.5G
101900-518EA 364.62 USD
101900-518 101900-520
3-Amino-N-benzyl-4-chloro-N-methylbenzenesulfonamide
3-Amino-N-benzyl-4-chloro-N-methylbenzenesulfonamide

Hazard: Irritant

Formula: C₁₄H₁₅ClN₂O₂S
MDL Number: MFCD12736530
CAS Number: 1036441-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR