1-[(3-Amino-4-chlorophenyl)sulfonyl]-4-piperidinol

Supplier: Matrix Scientific

052654-500MG 052654-2.5G
101900-502EA 364.62 USD
101900-502 101900-504
1-[(3-Amino-4-chlorophenyl)sulfonyl]-4-piperidinol
1-[(3-Amino-4-chlorophenyl)sulfonyl]-4-piperidinol

Hazard: Irritant

Formula: C₁₁H₁₅ClN₂O₃S
MDL Number: MFCD11118720
CAS Number: 1036500-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR