4-[(3-Amino-4-chlorophenyl)sulfonyl]-2-piperazinone

Supplier: Matrix Scientific

052653-500MG 052653-2.5G
101900-498EA 364.62 USD
101900-498 101900-500
4-[(3-Amino-4-chlorophenyl)sulfonyl]-2-piperazinone
4-[(3-Amino-4-chlorophenyl)sulfonyl]-2-piperazinone

Hazard: Irritant

Formula: C₁₀H₁₂ClN₃O₃S
MDL Number: MFCD11120109
CAS Number: 1041566-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR