2-Chloro-5-[(4-methyl-1-piperazinyl)sulfonyl]aniline

Supplier: Matrix Scientific

052650-500MG 052650-2.5G
101900-486EA 364.62 USD
101900-486 101900-488
2-Chloro-5-[(4-methyl-1-piperazinyl)sulfonyl]aniline
2-Chloro-5-[(4-methyl-1-piperazinyl)sulfonyl]aniline

Hazard: Irritant

Formula: C₁₁H₁₆ClN₃O₂S
MW: 289.79 g/mol
MDL Number: MFCD07323301
CAS Number: 848369-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR