2-Chloro-5-[(3-methyl-1-piperidinyl)sulfonyl]aniline

Supplier: Matrix Scientific

052648-500MG 052648-2.5G
101900-478EA 364.62 USD
101900-478 101900-480
2-Chloro-5-[(3-methyl-1-piperidinyl)sulfonyl]aniline
2-Chloro-5-[(3-methyl-1-piperidinyl)sulfonyl]aniline

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₂S
MW: 288.8 g/mol
MDL Number: MFCD11116282
CAS Number: 1036526-86-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR