2-Chloro-5-[(2-methyl-1-piperidinyl)sulfonyl]aniline

Supplier: Matrix Scientific

052647-500MG 052647-2.5G
101900-474EA 364.62 USD
101900-474 101900-476
2-Chloro-5-[(2-methyl-1-piperidinyl)sulfonyl]aniline
2-Chloro-5-[(2-methyl-1-piperidinyl)sulfonyl]aniline

Hazard: Irritant

Formula: C₁₂H₁₇ClN₂O₂S
MW: 288.8 g/mol
MDL Number: MFCD11117514
CAS Number: 1036470-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR