N,N-Diallyl-3-amino-4-chlorobenzenesulfonamide

Supplier: Matrix Scientific

052646-500MG 052646-2.5G
101900-470EA 364.62 USD
101900-470 101900-472
N,N-Diallyl-3-amino-4-chlorobenzenesulfonamide
N,N-Diallyl-3-amino-4-chlorobenzenesulfonamide

Hazard: Irritant

Formula: C₁₂H₁₅ClN₂O₂S
MW: 286.78 g/mol
MDL Number: MFCD13562330
CAS Number: 1219957-24-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR