3-Amino-4-chloro-N-ethyl-N-(2-hydroxyethyl)benzenesulfonamide

Supplier: Matrix Scientific

052645-500MG 052645-2.5G
101900-466EA 364.62 USD
101900-466 101900-468
3-Amino-4-chloro-N-ethyl-N-(2-hydroxyethyl)benzenesulfonamide
3-Amino-4-chloro-N-ethyl-N-(2-hydroxyethyl)benzenesulfonamide

Hazard: Irritant

Formula: C₁₀H₁₅ClN₂O₃S
MDL Number: MFCD12890902
CAS Number: 1184235-88-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR