3-Amino-N-butyl-4-chloro-N-methylbenzenesulfonamide

Supplier: Matrix Scientific

052644-500MG 052644-2.5G
101900-462EA 364.62 USD
101900-462 101900-464
3-Amino-N-butyl-4-chloro-N-methylbenzenesulfonamide
3-Amino-N-butyl-4-chloro-N-methylbenzenesulfonamide

Hazard: Irritant

Formula: C₁₁H₁₇ClN₂O₂S
MDL Number: MFCD11123782
CAS Number: 1040336-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR