2-Chloro-5-(4-morpholinylsulfonyl)aniline

Supplier: Matrix Scientific

052642-500MG 052642-2.5G
101900-454EA 364.62 USD
101900-454 101900-456
2-Chloro-5-(4-morpholinylsulfonyl)aniline
2-Chloro-5-(4-morpholinylsulfonyl)aniline

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O₃S
MDL Number: MFCD02707949
CAS Number: 99187-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR