3-Amino-4-chloro-N-(2-hydroxyethyl)-N-methylbenzenesulfonamide

Supplier: Matrix Scientific

052640-500MG 052640-2.5G
101900-446EA 364.62 USD
101900-446 101900-448
3-Amino-4-chloro-N-(2-hydroxyethyl)-N-methylbenzenesulfonamide
3-Amino-4-chloro-N-(2-hydroxyethyl)-N-methylbenzenesulfonamide

Hazard: Irritant

Formula: C₉H₁₃ClN₂O₃S
MDL Number: MFCD11642938
CAS Number: 1098363-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR