3-Amino-4-chloro-N,N-dimethylbenzenesulfonamide

Supplier: Matrix Scientific

052638-500MG 052638-2.5G
101900-438EA 364.62 USD
101900-438 101900-440
3-Amino-4-chloro-N,N-dimethylbenzenesulfonamide
3-Amino-4-chloro-N,N-dimethylbenzenesulfonamide

Hazard: Irritant

Formula: C₈H₁₁ClN₂O₂S
MDL Number: MFCD02704404
CAS Number: 100313-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR