2-[Benzyl(ethyl)amino]-N'-hydroxybenzenecarboximidamide

Supplier: Matrix Scientific

052636-500MG 052636-2.5G
101900-430EA 364.62 USD
101900-430 101900-432
2-[Benzyl(ethyl)amino]-N'-hydroxybenzenecarboximidamide
2-[Benzyl(ethyl)amino]-N'-hydroxybenzenecarboximidamide

Hazard: Irritant

Formula: C₁₆H₁₉N₃O
MDL Number: MFCD13562327
CAS Number: 1087792-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR