2-[3,4-Dihydro-1(2H)-quinolinyl]-N'-hydroxybenzenecarboximidamide

Supplier: Matrix Scientific

052635-500MG 052635-2.5G
101900-426EA 364.62 USD
101900-426 101900-428
2-[3,4-Dihydro-1(2H)-quinolinyl]-N'-hydroxybenzenecarboximidamide
2-[3,4-Dihydro-1(2H)-quinolinyl]-N'-hydroxybenzenecarboximidamide

Hazard: Irritant

Formula: C₁₆H₁₇N₃O
MDL Number: MFCD13562326
CAS Number: 1021235-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR