2-(2,3-Dihydro-1H-indol-1-yl)-N'-hydroxybenzenecarboximidamide

Supplier: Matrix Scientific

052634-500MG 052634-2.5G
101900-422EA 364.62 USD
101900-422 101900-424
2-(2,3-Dihydro-1H-indol-1-yl)-N'-hydroxybenzenecarboximidamide
2-(2,3-Dihydro-1H-indol-1-yl)-N'-hydroxybenzenecarboximidamide

Hazard: Irritant

Formula: C₁₅H₁₅N₃O
MDL Number: MFCD12452229
CAS Number: 1021235-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR