N'-Hydroxy-2-[4-(2-hydroxyethyl)-1-piperazinyl]benzenecarboximidamide

Supplier: Matrix Scientific

052632-500MG 052632-2.5G
101900-414EA 364.62 USD
101900-414 101900-416
N'-Hydroxy-2-[4-(2-hydroxyethyl)-1-piperazinyl]benzenecarboximidamide
N'-Hydroxy-2-[4-(2-hydroxyethyl)-1-piperazinyl]benzenecarboximidamide

Hazard: Irritant

Formula: C₁₃H₂₀N₄O₂
MW: 264.33 g/mol
MDL Number: MFCD12687324
CAS Number: 1158140-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR