2-[Cyclohexyl(ethyl)amino]-N'-hydroxybenzenecarboximidamide

Supplier: Matrix Scientific

052631-500MG 052631-2.5G
101900-410EA 364.62 USD
101900-410 101900-412
2-[Cyclohexyl(ethyl)amino]-N'-hydroxybenzenecarboximidamide
2-[Cyclohexyl(ethyl)amino]-N'-hydroxybenzenecarboximidamide

Hazard: Irritant

Formula: C₁₅H₂₃N₃O
MDL Number: MFCD12626786
CAS Number: 1021235-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR