2-[Benzyl(methyl)amino]-N'-hydroxybenzenecarboximidamide

Supplier: Matrix Scientific

052630-500MG 052630-2.5G
101900-406EA 364.62 USD
101900-406 101900-408
2-[Benzyl(methyl)amino]-N'-hydroxybenzenecarboximidamide
2-[Benzyl(methyl)amino]-N'-hydroxybenzenecarboximidamide

Hazard: Irritant

Formula: C₁₅H₁₇N₃O
MDL Number: MFCD12452225
CAS Number: 1021244-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR