2-[Cyclohexyl(methyl)amino]-N'-hydroxybenzenecarboximidamide

Supplier: Matrix Scientific

052628-500MG 052628-2.5G
101900-398EA 364.62 USD
101900-398 101900-400
2-[Cyclohexyl(methyl)amino]-N'-hydroxybenzenecarboximidamide
2-[Cyclohexyl(methyl)amino]-N'-hydroxybenzenecarboximidamide

Hazard: Irritant

Formula: C₁₄H₂₁N₃O
MDL Number: MFCD11528965
CAS Number: 1021245-02-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR