N'-Hydroxy-2-(4-hydroxy-1-piperidinyl)benzenecarboximidamide

Supplier: Matrix Scientific

052626-500MG 052626-2.5G
101900-390EA 364.62 USD
101900-390 101900-392
N'-Hydroxy-2-(4-hydroxy-1-piperidinyl)benzenecarboximidamide
N'-Hydroxy-2-(4-hydroxy-1-piperidinyl)benzenecarboximidamide

Hazard: Irritant

Formula: C₁₂H₁₇N₃O₂
MW: 235.29 g/mol
MDL Number: MFCD11155685
CAS Number: 1021235-49-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR