N'-Hydroxy-2-(3-oxo-1-piperazinyl)benzenecarboximidamide

Supplier: Matrix Scientific

052625-500MG 052625-2.5G
101900-386EA 364.62 USD
101900-386 101900-388
N'-Hydroxy-2-(3-oxo-1-piperazinyl)benzenecarboximidamide
N'-Hydroxy-2-(3-oxo-1-piperazinyl)benzenecarboximidamide

Hazard: Irritant

Formula: C₁₁H₁₄N₄O₂
MW: 234.26 g/mol
MDL Number: MFCD13562324
CAS Number: 1220040-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR