2-(4-Benzyl-1-piperazinyl)-N'-hydroxybenzenecarboximidamide

Supplier: Matrix Scientific

052623-500MG 052623-2.5G
101900-378EA 364.62 USD
101900-378 101900-380
2-(4-Benzyl-1-piperazinyl)-N'-hydroxybenzenecarboximidamide
2-(4-Benzyl-1-piperazinyl)-N'-hydroxybenzenecarboximidamide

Hazard: Irritant

Formula: C₁₈H₂₂N₄O
MDL Number: MFCD13562323
CAS Number: 1220040-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR