N'-Hydroxy-2-(4-methyl-1-piperazinyl)benzenecarboximidamide

Supplier: Matrix Scientific

052622-500MG 052622-2.5G
101900-374EA 364.62 USD
101900-374 101900-376
N'-Hydroxy-2-(4-methyl-1-piperazinyl)benzenecarboximidamide
N'-Hydroxy-2-(4-methyl-1-piperazinyl)benzenecarboximidamide

Hazard: Irritant

Formula: C₁₂H₁₈N₄O
MW: 234.3 g/mol
MDL Number: MFCD11155635
CAS Number: 1021244-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR