N'-Hydroxy-2-(3-methyl-1-piperidinyl)benzenecarboximidamide

Supplier: Matrix Scientific

052620-500MG 052620-2.5G
101900-366EA 364.62 USD
101900-366 101900-368
N'-Hydroxy-2-(3-methyl-1-piperidinyl)benzenecarboximidamide
N'-Hydroxy-2-(3-methyl-1-piperidinyl)benzenecarboximidamide

Hazard: Irritant

Formula: C₁₃H₁₉N₃O
MW: 233.32 g/mol
MDL Number: MFCD11155640
CAS Number: 1021244-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR