2-(Diallylamino)-N'-hydroxybenzenecarboximidamide

Supplier: Matrix Scientific

052618-500MG 052618-2.5G
101900-358EA 364.62 USD
101900-358 101900-360
2-(Diallylamino)-N'-hydroxybenzenecarboximidamide
2-(Diallylamino)-N'-hydroxybenzenecarboximidamide

Hazard: Irritant

Formula: C₁₃H₁₇N₃O
MDL Number: MFCD13562322
CAS Number: 1220040-45-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR