2-[Ethyl(2-hydroxyethyl)amino]-N'-hydroxybenzenecarboximidamide

Supplier: Matrix Scientific

052617-500MG 052617-2.5G
101900-354EA 364.62 USD
101900-354 101900-356
2-[Ethyl(2-hydroxyethyl)amino]-N'-hydroxybenzenecarboximidamide
2-[Ethyl(2-hydroxyethyl)amino]-N'-hydroxybenzenecarboximidamide

Hazard: Irritant

Formula: C₁₁H₁₇N₃O₂
MDL Number: MFCD12813632
CAS Number: 1216335-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR