2-[Butyl(methyl)amino]-N'-hydroxybenzenecarboximidamide

Supplier: Matrix Scientific

052616-500MG 052616-2.5G
101900-350EA 364.62 USD
101900-350 101900-352
2-[Butyl(methyl)amino]-N'-hydroxybenzenecarboximidamide
2-[Butyl(methyl)amino]-N'-hydroxybenzenecarboximidamide

Hazard: Irritant

Formula: C₁₂H₁₉N₃O
MDL Number: MFCD11155669
CAS Number: 1021244-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR