N'-Hydroxy-2-(3-hydroxy-1-pyrrolidinyl)benzenecarboximidamide

Supplier: Matrix Scientific

052615-500MG 052615-2.5G
101900-346EA 364.62 USD
101900-346 101900-348
N'-Hydroxy-2-(3-hydroxy-1-pyrrolidinyl)benzenecarboximidamide
N'-Hydroxy-2-(3-hydroxy-1-pyrrolidinyl)benzenecarboximidamide

Hazard: Irritant

Formula: C₁₁H₁₅N₃O₂
MW: 221.26 g/mol
MDL Number: MFCD13562321
CAS Number: 1220040-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR