N'-Hydroxy-2-(4-morpholinyl)benzenecarboximidamide

Supplier: Matrix Scientific

052614-500MG 052614-2.5G
101900-342EA 364.62 USD
101900-342 101900-344
N'-Hydroxy-2-(4-morpholinyl)benzenecarboximidamide
N'-Hydroxy-2-(4-morpholinyl)benzenecarboximidamide

Hazard: Irritant

Formula: C₁₁H₁₅N₃O₂
MDL Number: MFCD11155628
CAS Number: 1021243-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR