N'-Hydroxy-2-(1-piperidinyl)benzenecarboximidamide

Supplier: Matrix Scientific

052613-500MG 052613-2.5G
101900-338EA 364.62 USD
101900-338 101900-340
N'-Hydroxy-2-(1-piperidinyl)benzenecarboximidamide
N'-Hydroxy-2-(1-piperidinyl)benzenecarboximidamide

Hazard: Irritant

Formula: C₁₂H₁₇N₃O
MDL Number: MFCD05975041
CAS Number: 860784-50-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR