N'-Hydroxy-2-(1-pyrrolidinyl)benzenecarboximidamide

Supplier: Matrix Scientific

052610-500MG 052610-2.5G
101900-326EA 364.62 USD
101900-326 101900-328
N'-Hydroxy-2-(1-pyrrolidinyl)benzenecarboximidamide
N'-Hydroxy-2-(1-pyrrolidinyl)benzenecarboximidamide

Hazard: Irritant

Formula: C₁₁H₁₅N₃O
MW: 205.26 g/mol
MDL Number: MFCD11155625
CAS Number: 1021243-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR