N-[2-(Aminomethyl)phenyl]-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)amine

Supplier: Matrix Scientific

052608-500MG 052608-2.5G
101900-318EA 364.62 USD
101900-318 101900-320
N-[2-(Aminomethyl)phenyl]-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)amine
N-[2-(Aminomethyl)phenyl]-N-methyl-N-(tetrahydro-2H-pyran-4-ylmethyl)amine

Hazard: Irritant

Formula: C₁₄H₂₂N₂O
MDL Number: MFCD13562320
CAS Number: 1220027-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR