N-[2-(Aminomethyl)phenyl]-N-benzyl-N-ethylamine

Supplier: Matrix Scientific

052607-500MG 052607-2.5G
101900-314EA 364.62 USD
101900-314 101900-316
N-[2-(Aminomethyl)phenyl]-N-benzyl-N-ethylamine
N-[2-(Aminomethyl)phenyl]-N-benzyl-N-ethylamine

Hazard: Irritant

Formula: C₁₆H₂₀N₂
MW: 240.35 g/mol
MDL Number: MFCD11195370
CAS Number: 1039314-59-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR