{2-[3,4-Dihydro-1(2H)-quinolinyl]phenyl}methanamine

Supplier: Matrix Scientific

052606-500MG 052606-2.5G
101900-310EA 364.62 USD
101900-310 101900-312
{2-[3,4-Dihydro-1(2H)-quinolinyl]phenyl}methanamine
{2-[3,4-Dihydro-1(2H)-quinolinyl]phenyl}methanamine

Hazard: Irritant

Formula: C₁₆H₁₈N₂
MDL Number: MFCD08699755
CAS Number: 937599-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR