[2-(2,3-Dihydro-1H-indol-1-yl)phenyl]methanamine

Supplier: Matrix Scientific

052605-500MG 052605-2.5G
101900-306EA 364.62 USD
101900-306 101900-308
[2-(2,3-Dihydro-1H-indol-1-yl)phenyl]methanamine
[2-(2,3-Dihydro-1H-indol-1-yl)phenyl]methanamine

Hazard: Irritant

Formula: C₁₅H₁₆N₂
MDL Number: MFCD08699754
CAS Number: 92083-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR