2-{4-[2-(Aminomethyl)phenyl]-1-piperazinyl}-1-ethanol
Supplier: Matrix Scientific
052603-500MG
052603-2.5G
101900-298EA
364.62
USD
101900-298
101900-300
2-{4-[2-(Aminomethyl)phenyl]-1-piperazinyl}-1-ethanol
2-{4-[2-(Aminomethyl)phenyl]-1-piperazinyl}-1-ethanol
Hazard: Irritant
Formula:
C₁₃H₂₁N₃O |
MDL Number:
MFCD10033350 CAS Number: 1018656-79-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...