2-(Aminomethyl)-N-cyclohexyl-N-ethylaniline

Supplier: Matrix Scientific

052602-500MG 052602-2.5G
101900-294EA 364.62 USD
101900-294 101900-296
2-(Aminomethyl)-N-cyclohexyl-N-ethylaniline
2-(Aminomethyl)-N-cyclohexyl-N-ethylaniline

Hazard: Irritant

Formula: C₁₅H₂₄N₂
MDL Number: MFCD11155606
CAS Number: 1021237-25-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR