[2-(2-Ethyl-1-piperidinyl)phenyl]methanamine

Supplier: Matrix Scientific

052600-500MG 052600-2.5G
101900-286EA 364.62 USD
101900-286 101900-288
[2-(2-Ethyl-1-piperidinyl)phenyl]methanamine
[2-(2-Ethyl-1-piperidinyl)phenyl]methanamine

Hazard: Irritant

Formula: C₁₄H₂₂N₂
MDL Number: MFCD11624782
CAS Number: 1094828-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR