2-(Aminomethyl)-N-cyclohexyl-N-methylaniline

Supplier: Matrix Scientific

052599-500MG 052599-2.5G
101900-282EA 364.62 USD
101900-282 101900-284
2-(Aminomethyl)-N-cyclohexyl-N-methylaniline
2-(Aminomethyl)-N-cyclohexyl-N-methylaniline

Hazard: Irritant

Formula: C₁₄H₂₂N₂
MDL Number: MFCD07365241
CAS Number: 893752-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR