4-[2-(Aminomethyl)phenyl]-2-piperazinone

Supplier: Matrix Scientific

052596-500MG 052596-2.5G
101900-270EA 364.62 USD
101900-270 101900-272
4-[2-(Aminomethyl)phenyl]-2-piperazinone
4-[2-(Aminomethyl)phenyl]-2-piperazinone

Hazard: Irritant

Formula: C₁₁H₁₅N₃O
MDL Number: MFCD11155590
CAS Number: 1021236-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR