[2-(1-Azepanyl)phenyl]methanamine

Supplier: Matrix Scientific

052595-500MG 052595-2.5G
101900-266EA 186.15 USD
101900-266 101900-268
[2-(1-Azepanyl)phenyl]methanamine
[2-(1-Azepanyl)phenyl]methanamine

Hazard: Irritant

Formula: C₁₃H₂₀N₂
MDL Number: MFCD06213555
CAS Number: 72752-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR