[2-(4-Benzyl-1-piperazinyl)phenyl]methanamine

Supplier: Matrix Scientific

052594-500MG 052594-2.5G
101900-262EA 364.62 USD
101900-262 101900-264
[2-(4-Benzyl-1-piperazinyl)phenyl]methanamine
[2-(4-Benzyl-1-piperazinyl)phenyl]methanamine

Hazard: Irritant

Formula: C₁₈H₂₃N₃
MDL Number: MFCD07365245
CAS Number: 321908-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR