[2-(4-Methyl-1-piperazinyl)phenyl]methanamine

Supplier: Matrix Scientific

052593-500MG 052593-2.5G
101900-258EA 364.62 USD
101900-258 101900-260
[2-(4-Methyl-1-piperazinyl)phenyl]methanamine
[2-(4-Methyl-1-piperazinyl)phenyl]methanamine

Hazard: Irritant

Formula: C₁₂H₁₉N₃
MW: 205.31 g/mol
MDL Number: MFCD04970994
CAS Number: 655256-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR