[2-(2-Methyl-1-piperidinyl)phenyl]methanamine

Supplier: Matrix Scientific

052592-500MG 052592-2.5G
101900-254EA 364.62 USD
101900-254 101900-256
[2-(2-Methyl-1-piperidinyl)phenyl]methanamine
[2-(2-Methyl-1-piperidinyl)phenyl]methanamine

Hazard: Irritant

Formula: C₁₃H₂₀N₂
MDL Number: MFCD04971083
CAS Number: 869943-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR