N,N-Diallyl-2-(aminomethyl)aniline

Supplier: Matrix Scientific

052591-500MG 052591-2.5G
101900-250EA 364.62 USD
101900-250 101900-252
N,N-Diallyl-2-(aminomethyl)aniline
N,N-Diallyl-2-(aminomethyl)aniline

Hazard: Irritant

Formula: C₁₃H₁₈N₂
MDL Number: MFCD13562319
CAS Number: 1220034-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR