2-(Aminomethyl)-N-butyl-N-methylaniline

Supplier: Matrix Scientific

052589-500MG 052589-2.5G
101900-242EA 364.62 USD
101900-242 101900-244
2-(Aminomethyl)-N-butyl-N-methylaniline
2-(Aminomethyl)-N-butyl-N-methylaniline

Hazard: Irritant

Formula: C₁₂H₂₀N₂
MDL Number: MFCD11155633
CAS Number: 1020988-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR